Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oas1h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009491.1 | NP_001009491.1 | 4 | 913 | ||
XM_008769247.1 | XP_008767469.1 | 4 | 990 | |||
GenBank mRNA | AY196701.1 | - | 4 | 913 |