Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vom1r20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009506.1 | NP_001009506.1 | 1 | 15 | ||
GenBank mRNA | AY510344.1 | - | 1 | 15 |