Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vom1r74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008911.1 | NP_001008911.1 | 1 | 768 | ||
GenBank mRNA | AY510285.1 | - | 1 | 768 |
Gene Symbol: LOC100909399 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749767.1 | XP_003749815.1 | 1 | 768 |