Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vom1r23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008941.1 | NP_001008941.1 | 1 | 930 | ||
GenBank mRNA | AY510359.1 | - | 1 | 930 |
Gene Symbol: Vom1r-ps112 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008758822.1 | XP_008757044.1 | 1 | 991 |