Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gsta3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009920.2 | NP_001009920.1 | 5 | 878 | ||
NM_001159739.2 | NP_001153211.1 | 6 | 848 | |||
NM_001287025.1 | NP_001273954.1 | 5 | 740 | |||
GenBank mRNA | BQ201014.1 | - | 1 | 280 | ||
DY573045.1 | - | 4 | 573 | |||
FQ210706.1 | - | 5 | 707 | |||
FQ218526.1 | - | 6 | 721 | |||
FQ218726.1 | - | 6 | 715 | |||
FQ219557.1 | - | 6 | 715 | |||
FQ219615.1 | - | 6 | 848 | |||
FQ219704.1 | - | 6 | 715 | |||
FQ231428.1 | - | 6 | 720 | |||
S72506.1 | - | 5 | 726 | |||
S82820.1 | - | 5 | 878 | |||
X78847.1 | - | 5 | 726 |