Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xkr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012228.1 | NP_001012228.1 | 5 | 1415 | ||
GenBank mRNA | AY534259.1 | - | 5 | 1415 |