Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pik3r6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081444.1 | NP_001074913.1 | 15, 16 | 1892 | ||
XM_008767851.1 | XP_008766073.1 | 18, 19 | 2358 | |||
GenBank mRNA | EF207569.1 | - | 15, 16 | 1892 |