Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp652 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080207.1 | NP_001073676.1 | 1 | 954 | ||
GenBank mRNA | BC129097.1 | - | 1 | 954 | 8365300 |