Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Med1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134361.1 | NP_001127833.1 | 5 | 396 | ||
XM_006247398.2 | XP_006247460.1 | 5 | 546 | |||
XM_006247399.2 | XP_006247461.1 | 6 | 621 | |||
XM_006247401.2 | XP_006247463.1 | 5 | 545 | |||
XM_008768105.1 | XP_008766327.1 | 5 | 1176 |