Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGD1565033 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109050.1 | NP_001102520.1 | 3 | 362 | ||
XM_006247636.1 | XP_006247698.1 | 4 | 1169 | |||
XM_006247637.1 | XP_006247699.1 | 4 | 1166 | |||
XM_006247638.1 | XP_006247700.1 | 5 | 1046 | |||
XM_006247639.2 | XP_006247701.1 | 4 | 1227 | |||
XM_006247640.2 | XP_006247702.1 | 4 | 1199 | |||
XM_006247641.2 | XP_006247703.1 | 3 | 1107 | |||
XM_008768383.1 | XP_008766605.1 | 5 | 824 | |||
GenBank mRNA | BC158779.1 | - | 2 | 1178 | 7134339 |