Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751133.2 | XP_003751181.1 | 2, 3 | 295 | ||
XM_006248968.2 | XP_006249030.1 | 2, 3 | 250 |