Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109081.1 | NP_001102551.1 | 6 | 648 | ||
GenBank mRNA | FQ226610.1 | - | 642 | |||
FQ227016.1 | - | 642 |