Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coq2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044255.1 | NP_001037720.1 | 3 | 523 | ||
XM_006250675.2 | XP_006250737.1 | 3 | 555 | |||
GenBank mRNA | BC099827.1 | - | 3 | 523 | 7934175 |