Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrps24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077659.1 | NP_001071127.1 | 4 | 490 | ||
GenBank mRNA | AF034241.1 | - | 1 | 33 | ||
BC157807.1 | - | 4 | 486 | 8375708 | ||
BP503351.1 | - | 4 | 495 | |||
DY319747.1 | - | 4 | 486 | |||
FQ227686.1 | - | 4 | 508 |