Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsc22d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109912.1 | NP_001103382.1 | 3 | 3334 | ||
NM_013043.2 | NP_037175.1 | 3 | 462 | |||
XM_006252339.1 | XP_006252401.1 | 4 | 3206 | |||
GenBank mRNA | BC059146.1 | - | 3 | 473 | ||
BC127547.1 | - | 3 | 454 | |||
FQ229551.1 | - | 1 | 21 | |||
L25785.1 | - | 3 | 455 |