Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ldb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110490.2 | NP_001103960.1 | 6 | 508 | ||
NM_001277158.1 | NP_001264087.1 | 6 | 508 | |||
NM_001277159.1 | NP_001264088.1 | 6 | 498 | |||
NM_001277166.1 | NP_001264095.1 | 6 | 498 | |||
XM_006252640.2 | XP_006252702.1 | 6 | 554 | |||
XM_006252644.2 | XP_006252706.1 | 6 | 554 | |||
XM_006252647.1 | XP_006252709.1 | 6 | 554 | |||
GenBank mRNA | BC083721.1 | - | 5 | 446 | 7190925 | |
FM122871.1 | - | 4 | 264 | |||
FN804937.1 | - | 6 | 466 | |||
FQ214774.1 | - | 5 | 352 | |||
FQ215617.1 | - | 6 | 494 | |||
FQ216341.1 | - | 6 | 504 | |||
FQ216348.1 | - | 6 | 504 | |||
FQ217970.1 | - | 6 | 504 | |||
FQ223559.1 | - | 6 | 504 |