Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mef2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017507.1 | NP_001017507.1 | 4 | 391 | ||
XM_006252927.2 | XP_006252989.1 | 4 | 711 | |||
GenBank mRNA | BC079361.1 | - | 4 | 391 | 7109152 |