Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aaed1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109114.1 | NP_001102584.1 | 4 | 414 | ||
XM_006253495.2 | XP_006253557.1 | 4 | 414 | |||
XM_006253497.2 | XP_006253559.1 | 4 | 418 | |||
XM_008771445.1 | XP_008769667.1 | 4 | 423 |