Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGD1560860 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170421.1 | NP_001163892.1 | 6 | 584 | ||
XM_008771753.1 | XP_008769975.1 | 6 | 792 | |||
XM_008771754.1 | XP_008769976.1 | 7 | 616 | |||
XM_008771755.1 | XP_008769977.1 | 6 | 588 | |||
XM_008771756.1 | XP_008769978.1 | 6 | 621 | |||
XM_008771757.1 | XP_008769979.1 | 6 | 975 | |||
XM_008771758.1 | XP_008769980.1 | 6 | 741 | |||
XM_008771759.1 | XP_008769981.1 | 6 | 530 | |||
XM_008771760.1 | XP_008769982.1 | 4 | 803 | |||
XM_008771761.1 | XP_008769983.1 | 5 | 423 | |||
GenBank mRNA | BC158594.1 | - | 6 | 584 | 7111099 | |
FQ225186.1 | - | 2 | 266 | |||
FQ231877.1 | - | 2 | 266 |
Gene Symbol: LOC103690053 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008771792.1 | XP_008770014.1 | 4 | 793 |