Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtcp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_574098.4 | XP_574098.2 | 3 | 384 |
Gene Symbol: LOC102546764 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006254392.1 | XP_006254454.1 | 1 | 17 |