Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ces2h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044258.2 | NP_001037723.1 | 10 | 1425 | ||
XM_008772523.1 | XP_008770745.1 | 10 | 1444 | |||
XM_008772524.1 | XP_008770746.1 | 11 | 1567 | |||
GenBank mRNA | BC107806.1 | - | 10 | 1395 | 7368719 |