Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wtap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113542.1 | NP_001107014.1 | 6 | 629 | ||
NM_001113543.1 | NP_001107015.1 | 6 | 629 | |||
NM_001113544.1 | NP_001107016.1 | 6 | 563 | |||
XM_006227892.2 | XP_006227954.1 | 6 | 609 | |||
XM_006227893.2 | XP_006227955.1 | 6 | 787 | |||
GenBank mRNA | CK478322.1 | - | 6 | 629 | ||
CV108433.1 | - | 6 | 626 | |||
CX570823.1 | - | 2 | 275 | |||
FQ228682.1 | - | 6 | 668 |