Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGD1564380 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008758911.1 | XP_008757133.1 | 6 | 927 | ||
XM_008758912.1 | XP_008757134.1 | 6 | 935 | |||
XM_008758913.1 | XP_008757135.1 | 6 | 788 | |||
XM_008758914.1 | XP_008757136.1 | 6 | 930 |
Gene Symbol: LOC102553892 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008759075.1 | XP_008757297.1 | 5 | 602 |
Gene Symbol: RGD1565655 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177686.1 | NP_001171157.2 | 5 | 1140 | ||
XM_008758968.1 | XP_008757190.1 | 602 | ||||
GenBank mRNA | BC158626.1 | - | 5 | 1140 | 7455240 |