Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dmrtc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109140.1 | NP_001102610.1 | 2 | 136 |
Gene Symbol: LOC100910371 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006244579.2 | XP_006244641.1 | 2 | 190 | ||
XM_008766899.1 | XP_008765121.1 | 1 | 789 | |||
XM_008766900.1 | XP_008765122.1 | 1 | 789 |