Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdv3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006243659.2 | XP_006243721.1 | 5 | 1111 | ||
XM_008766529.1 | XP_008764751.1 | 5 | 1740 |
Gene Symbol: LOC499235 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_574528.5 | XP_574528.1 | 2 | 435 | ||
GenBank mRNA | AY539892.1 | - | 2 | 435 |