Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam24a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109158.1 | NP_001102628.1 | 3 | 370 | ||
XM_006230419.2 | XP_006230481.1 | 5 | 737 | |||
XM_008759917.1 | XP_008758139.1 | 3 | 289 | |||
GenBank mRNA | BC161873.1 | - | 5 | 678 | 7113177 | |
BC166439.1 | - | 3 | 297 | 7453416 |