Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Icoslg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006256260.2 | XP_006256322.1 | 4 | 849 | ||
XM_006256261.2 | XP_006256323.1 | 4 | 846 | |||
XM_006256262.2 | XP_006256324.1 | 4 | 848 | |||
XM_006256264.2 | XP_006256326.1 | 4 | 847 |