Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lhfp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109183.1 | NP_001102653.1 | 4 | 923 | ||
GenBank mRNA | BC091355.1 | - | 3 | 658 | 7318141 |