Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il36g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113790.1 | NP_001107262.1 | 2, 3 | 193 | ||
GenBank mRNA | BC158611.1 | - | 2, 3 | 193 | 7384725 |