Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Man1b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109196.1 | NP_001102666.1 | 12 | 1735 | ||
GenBank mRNA | BC166449.1 | - | 12 | 1735 | ||
FQ210932.1 | - | 9 | 1390 |