Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slx4ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271320.1 | NP_001258249.1 | 3 | 231 | ||
XM_008762255.1 | XP_008760477.1 | 3 | 105 | |||
XM_008762256.1 | XP_008760478.1 | 4 | 503 | |||
XM_008762257.1 | XP_008760479.1 | 3 | 365 | |||
XM_008762258.1 | XP_008760480.1 | 3 | 102 | |||
XM_008762261.1 | XP_008760483.1 | 3 | 398 |