Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gins1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109207.2 | NP_001102677.2 | 1 | 172 | ||
GenBank mRNA | BC166989.1 | - | 1 | 172 | 9098732 |