Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gins1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109207.2 | NP_001102677.2 | 2 | 208 | ||
GenBank mRNA | BC166989.1 | - | 2 | 208 | 9098732 |