Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bpifb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109208.1 | NP_001102678.1 | 8 | 819 | ||
XM_006235352.2 | XP_006235414.1 | 7 | 721 | |||
GenBank mRNA | X60658.1 | - | 8 | 785 |