Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sla2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172118.1 | NP_001165589.1 | 4, 5 | 382 | ||
XM_008762374.1 | XP_008760596.1 | 6, 7 | 2059 |