Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dbndd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047111.2 | NP_001040576.1 | 2 | 470 | ||
XM_006235610.1 | XP_006235672.1 | 3 | 533 | |||
XM_006235612.1 | XP_006235674.1 | 3 | 289 | |||
XM_006235614.2 | XP_006235676.1 | 3 | 432 | |||
XM_006235615.2 | XP_006235677.1 | 3 | 378 | |||
GenBank mRNA | AY113696.1 | - | 2 | 348 | ||
BC158561.1 | - | 2 | 470 | 7387488 | ||
FQ213987.1 | - | 2 | 382 | |||
FQ220039.1 | - | 3 | 358 |