Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apcdd1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006235711.2 | XP_006235773.1 | 4 | 1546 | ||
XM_008762580.1 | XP_008760802.1 | 4 | 3628 | |||
GenBank mRNA | BC081904.1 | - | 1386 | 7110948 |