Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam221a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025771.2 | NP_001020942.1 | 7 | 916 | ||
XM_006236467.2 | XP_006236529.1 | 7 | 889 | |||
XM_006236468.2 | XP_006236530.1 | 6 | 900 | |||
GenBank mRNA | BC097434.1 | - | 7 | 917 | 7455430 |