Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109245.1 | NP_001102715.1 | 9 | 1017 | ||
XM_006236777.2 | XP_006236839.1 | 8 | 832 | |||
XM_006236778.2 | XP_006236840.1 | 9 | 936 | |||
XM_006236780.2 | XP_006236842.1 | 9 | 912 | |||
XM_006236781.2 | XP_006236843.1 | 9 | 2358 |
Gene Symbol: LOC103692166 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008763039.1 | XP_008761261.1 | 1425 | |||
XM_008763040.1 | XP_008761262.1 | 971 | ||||
XM_008763041.1 | XP_008761263.1 | 1043 | ||||
XM_008763042.1 | XP_008761264.1 | 939 |