Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Emx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008763096.1 | XP_008761318.1 | 1 | 874 | ||
XM_575584.5 | XP_575584.5 | 1 | 874 | |||
GenBank mRNA | FQ212435.1 | - | 1 | 813 |