Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGD1560289 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134623.2 | NP_001128095.2 | 11 | 1840 | ||
XM_006236917.2 | XP_006236979.1 | 11 | 3156 | |||
XM_008763127.1 | XP_008761349.1 | 10 | 4881 | |||
XM_008763128.1 | XP_008761350.1 | 11 | 3461 | |||
XM_008763129.1 | XP_008761351.1 | 9 | 4872 | |||
XM_008763130.1 | XP_008761352.1 | 10 | 4882 |