Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam110b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024341.1 | NP_001019512.1 | 1 | 1236 | ||
XM_006237839.2 | XP_006237901.1 | 3 | 2529 | |||
XM_006237840.2 | XP_006237902.1 | 4 | 1757 | |||
XM_006237841.2 | XP_006237903.1 | 3 | 1670 | |||
XM_006237843.2 | XP_006237905.1 | 2 | 6860 | |||
XM_008763551.1 | XP_008761773.1 | 2 | 7634 | |||
XM_008763552.1 | XP_008761774.1 | 3 | 5389 | |||
XM_008763553.1 | XP_008761775.1 | 4 | 5088 | |||
XM_008763554.1 | XP_008761776.1 | 4 | 5338 | |||
XM_008763555.1 | XP_008761777.1 | 2 | 2442 | |||
GenBank mRNA | BC091289.1 | - | 1 | 1236 | 7306431 |