Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134969.1 | NP_001128441.1 | 13 | 1591 | ||
XM_006237976.2 | XP_006238038.1 | 13 | 1520 | |||
XM_006237977.2 | XP_006238039.1 | 13 | 1492 | |||
XM_006237978.2 | XP_006238040.1 | 13 | 1950 | |||
XM_006237981.2 | XP_006238043.1 | 9 | 946 | |||
XM_008763638.1 | XP_008761860.1 | 13 | 1950 | |||
XM_008763639.1 | XP_008761861.1 | 12 | 1393 | |||
XM_008763640.1 | XP_008761862.1 | 13 | 1492 | |||
GenBank mRNA | BC168675.1 | - | 14 | 1527 | 7939387 |