Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134969.1 | NP_001128441.1 | 5 | 727 | ||
XM_006237976.2 | XP_006238038.1 | 5 | 656 | |||
XM_006237977.2 | XP_006238039.1 | 5 | 628 | |||
XM_006237978.2 | XP_006238040.1 | 5 | 1086 | |||
XM_008763638.1 | XP_008761860.1 | 5 | 1086 | |||
XM_008763639.1 | XP_008761861.1 | 5 | 628 | |||
XM_008763640.1 | XP_008761862.1 | 5 | 628 | |||
GenBank mRNA | BC168675.1 | - | 6 | 663 | 7939387 |