Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam110d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109270.1 | NP_001102740.1 | 2 | 262 | ||
XM_006239122.2 | XP_006239184.1 | 3 | 371 |