Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Camta1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195559.1 | NP_001182488.1 | 188 | |||
NM_001195560.1 | NP_001182489.1 | 3 | 188 | |||
GenBank mRNA | BC106025.1 | - | 3 | 188 | ||
BF290447.1 | - | 2 | 40 |