Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnfrsf18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024349.2 | NP_001019520.2 | 1 | 163 | ||
XM_008764373.1 | XP_008762595.1 | 1 | 1418 | |||
XM_008764374.1 | XP_008762596.1 | 2 | 1336 | |||
GenBank mRNA | BC088265.1 | - | 1 | 164 | 7377562 | |
FM088311.1 | - | 1 | 163 |