Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGD1566220 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006240360.2 | XP_006240422.1 | 1 | 595 | ||
XM_006240361.2 | XP_006240423.1 | 2 | 652 | |||
XM_006240362.2 | XP_006240424.1 | 2 | 661 | |||
XM_006240363.2 | XP_006240425.2 | 2 | 741 |