Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccnk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109672.1 | NP_001103142.1 | 6 | 571 | ||
XM_006240549.2 | XP_006240611.1 | 7 | 814 | |||
XM_006240551.2 | XP_006240613.1 | 5 | 816 | |||
GenBank mRNA | BC129121.1 | - | 6 | 571 | 7125896 |