Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp839 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006240613.2 | XP_006240675.1 | 2 | 1993 | ||
XM_006240614.2 | XP_006240676.1 | 2 | 1993 | |||
XM_006240615.2 | XP_006240677.1 | 2 | 1488 | |||
XM_006240616.2 | XP_006240678.2 | 2 | 2197 | |||
XM_008764973.1 | XP_008763195.1 | 2 | 2198 |